α,α,α-Trifluoro-o-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-(Trifluoromethyl)benzoic acid

001412-25G 001412-100G
FLUO001412-25GEA 0 GBP
FLUO001412-25G FLUO001412-100G
α,α,α-Trifluoro-o-toluic acid
α,α,α-Trifluoro-o-toluic acid
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 247
Melting Pt: 110
MDL Number: MFCD00002476
CAS Number: 433-97-6
EINECS: 207-093-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR