3-(Trifluoromethyl)benzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007583-10G 007583-50G
FLUO007583-10GEA 0 GBP
FLUO007583-10G FLUO007583-50G
3-(Trifluoromethyl)benzamide
3-(Trifluoromethyl)benzamide
Formula: C₈H₆F₃NO
MDL Number: MFCD00014802
CAS Number: 1801-10-1
EINECS: 217-289-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR