2-(Trifluoromethyl)benzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007582-10G 007582-50G
FLUO007582-10GEA 0 GBP
FLUO007582-10G FLUO007582-50G
2-(Trifluoromethyl)benzamide
2-(Trifluoromethyl)benzamide
Formula: C₈H₆F₃NO
MDL Number: MFCD00014798
CAS Number: 360-64-5
EINECS: 206-637-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR