α,α,α-Trifluoro-p-toluoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001413-25G
001413-100G
FLUO001413-25GEA
0
GBP
FLUO001413-25G
FLUO001413-100G
α,α,α-Trifluoro-p-toluoyl chloride
α,α,α-Trifluoro-p-toluoyl chloride
Formula:
C₈H₄ClF₃O Boiling Pt: 188…190 °C (1013 hPa) Melting Pt: –3 °C Density: 1.4035 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000694 CAS Number: 329-15-7 EINECS: 206-342-9 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...