2-Fluoro-6-(trifluoromethyl)benzoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006405-1G
006405-5G
006405-25G
FLUO006405-1GEA
0
GBP
FLUO006405-1G
FLUO006405-5G
FLUO006405-25G
2-Fluoro-6-(trifluoromethyl)benzoyl chloride
2-Fluoro-6-(trifluoromethyl)benzoyl chloride
Formula:
C₈H₃ClF₄O Boiling Pt: 193…195 °C (1013 hPa) Density: 1.465 g/cm³ (20 °C) |
MDL Number:
MFCD00061160 CAS Number: 109227-12-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...