α,α,α-Trifluoro-o-toluoyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(Trifluoromethyl)benzoyl chloride, 2-(Trifluoromethyl)benzene-1-carbonyl chloride
007597-10G
007597-50G
007597-100G
FLUO007597-10GEA
0
GBP
FLUO007597-10G
FLUO007597-50G
FLUO007597-100G
α,α,α-Trifluoro-o-toluoyl chloride
α,α,α-Trifluoro-o-toluoyl chloride
Formula:
C₈H₄ClF₃O Boiling Pt: 185…186 °C (1013 hPa) Melting Pt: –22 °C Density: 1.4518 g/cm³ (20 °C) |
MDL Number:
MFCD00000667 CAS Number: 312-94-7 EINECS: 206-233-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...