4'-Fluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone

001621-250G 001621-1KG 001621-100G
FLUO001621-250GEA 0 GBP
FLUO001621-250G FLUO001621-1KG FLUO001621-100G
4'-Fluoroacetophenone
4'-Fluoroacetophenone
Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 196
Density: 1.138 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000354
CAS Number: 403-42-9
EINECS: 206-960-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR