3'-Fluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-(3-Fluorophenyl)ethanone, m-Fluoroacetophenone

001467-25G 001467-100G
FLUO001467-25GEA 0 GBP
FLUO001467-25G FLUO001467-100G
3'-Fluoroacetophenone
3'-Fluoroacetophenone
Formula: C₈H₇FO
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1,12 g/cm³ (20 °C)
MDL Number: MFCD00000338
CAS Number: 455-36-7
EINECS: 207-245-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR