3',4',5'-Trifluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007953-1G 007953-5G
FLUO007953-1GEA 0 GBP
FLUO007953-1G FLUO007953-5G
3',4',5'-Trifluoroacetophenone
3',4',5'-Trifluoroacetophenone
Formula: C₈H₅F₃O
MDL Number: MFCD00083533
CAS Number: 220141-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR