2',3',4'-Trifluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007469-5G 007469-1G
FLUO007469-5GEA 0 GBP
FLUO007469-5G FLUO007469-1G
2',3',4'-Trifluoroacetophenone
2',3',4'-Trifluoroacetophenone
Formula: C₈H₅F₃O
MDL Number: MFCD00061231
CAS Number: 243448-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR