3',5'-Difluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(3,5-Difluorophenyl)ethan-1-one

002108-5G 002108-25G 002108-100G
FLUO002108-5GEA 0 GBP
FLUO002108-5G FLUO002108-25G FLUO002108-100G
3',5'-Difluoroacetophenone
3',5'-Difluoroacetophenone
Formula: C₈H₆F₂O
MDL Number: MFCD00042489
CAS Number: 123577-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR