2',4'-Difluoroacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-(2,4-Difluorophenyl)ethan-1-one
001914-25G
001914-100G
001914-250G
FLUO001914-25GEA
0
GBP
FLUO001914-25G
FLUO001914-100G
FLUO001914-250G
2',4'-Difluoroacetophenone
2',4'-Difluoroacetophenone
Formula:
C₈H₆F₂O Boiling Pt: 80 °C (1013 hPa) Density: 1,23 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151261 CAS Number: 364-83-0 EINECS: 206-667-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...