3',4'-Difluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(3,4-Difluorophenyl)ethan-1-one, 1-(3,4-Difluorophenyl)ethanone

002078-25G 002078-100G
FLUO002078-25GEA 0 GBP
FLUO002078-25G FLUO002078-100G
3',4'-Difluoroacetophenone
3',4'-Difluoroacetophenone
Formula: C₈H₆F₂O
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1,24 g/cm³ (20 °C)
MDL Number: MFCD00009891
CAS Number: 369-33-5
EINECS: 206-717-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR