α,α,α-Trifluoroacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone
001244-100G
001244-25G
001244-5G
FLUO001244-100GEA
0
GBP
FLUO001244-100G
FLUO001244-25G
FLUO001244-5G
α,α,α-Trifluoroacetophenone
α,α,α-Trifluoroacetophenone
Formula:
C₈H₅F₃O MW: 174.12 g/mol Boiling Pt: 165 °C (1013 hPa) Melting Pt: –40 °C Density: 1.2799 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000420 CAS Number: 434-45-7 EINECS: 207-103-1 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...