(4-Fluorophenyl)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluorobenzyl cyanide, 4-Fluorophenylacetonitrile

001189.25G 001189.100G
FLUO001189.25GEA 0 GBP
FLUO001189.25G FLUO001189.100G
(4-Fluorophenyl)acetonitrile
(4-Fluorophenyl)acetonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.136 g/cm³ (20 °C)
MDL Number: MFCD00001917
CAS Number: 459-22-3
EINECS: 207-286-8
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR