(3,4-Difluorophenyl)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004853-5G 004853-25G
FLUO004853-5GEA 0 GBP
FLUO004853-5G FLUO004853-25G
(3,4-Difluorophenyl)acetonitrile
(3,4-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
Boiling Pt: 110…120 °C (13 mmHg)
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00009974
CAS Number: 658-99-1
EINECS: 211-528-8
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR