(3,4,5-Trifluorophenyl)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007701-1G 007701-5G 007701-2.5G
FLUO007701-1GEA 0 GBP
FLUO007701-1G FLUO007701-5G FLUO007701-2.5G
(3,4,5-Trifluorophenyl)acetonitrile
(3,4,5-Trifluorophenyl)acetonitrile
Formula: C₈H₄F₃N
MDL Number: MFCD00083531
CAS Number: 220228-03-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR