(2,6-Difluorophenyl)acetonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004852-1G
004852-5G
004852-10G
004852-25G
FLUO004852-1GEA
0
GBP
FLUO004852-1G
FLUO004852-5G
FLUO004852-10G
FLUO004852-25G
(2,6-Difluorophenyl)acetonitrile
(2,6-Difluorophenyl)acetonitrile
Formula:
C₈H₅F₂N Boiling Pt: 109 °C (27 mmHg) Density: 1.2811 g/cm³ (20 °C) |
MDL Number:
MFCD00009973 CAS Number: 654-01-3 EINECS: 211-504-7 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...