(2,5-Difluorophenyl)acetonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,5-Difluorophenylacetonitrile, 2,5-Difluorobenzyl cyanide
004851-1G
004851-5G
004851-25G
FLUO004851-1GEA
0
GBP
FLUO004851-1G
FLUO004851-5G
FLUO004851-25G
(2,5-Difluorophenyl)acetonitrile
(2,5-Difluorophenyl)acetonitrile
Formula:
C₈H₅F₂N Boiling Pt: 90 °C (5 mmHg) Density: 1.23 g/cm³ (20 °C) |
MDL Number:
MFCD00009972 CAS Number: 69584-87-8 EINECS: 274-051-4 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...