(2,4-Difluorophenyl)acetonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004850-1G
004850-5G
FLUO004850-1GEA
0
GBP
FLUO004850-1G
FLUO004850-5G
(2,4-Difluorophenyl)acetonitrile
(2,4-Difluorophenyl)acetonitrile
Formula:
C₈H₅F₂N Boiling Pt: 90 °C (0.3 mmHg) Density: 1.2503 g/cm³ (15 °C) |
MDL Number:
MFCD00009971 CAS Number: 656-35-9 EINECS: 211-514-1 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...