(2,3-Difluorophenyl)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004849-5G 004849.10G 004849-1G
FLUO004849-5GEA 0 GBP
FLUO004849-5G FLUO004849.10G FLUO004849-1G
(2,3-Difluorophenyl)acetonitrile
(2,3-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 24 °C
MDL Number: MFCD00061277
CAS Number: 145689-34-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR