(2,3,6-Trifluorophenyl)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007698-5G 007698-1G
FLUO007698-5GEA 0 GBP
FLUO007698-5G FLUO007698-1G
(2,3,6-Trifluorophenyl)acetonitrile
(2,3,6-Trifluorophenyl)acetonitrile
Formula: C₈H₄F₃N
MDL Number: MFCD00061219
CAS Number: 114152-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR