(Pentafluorophenyl)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002404-5G 002404-10G
FLUO002404-5GEA 0 GBP
FLUO002404-5G FLUO002404-10G
(Pentafluorophenyl)acetonitrile
(Pentafluorophenyl)acetonitrile
Formula: (C₆F₅)CH₂CN
Boiling Pt: 107 °C (1013 hPa)
Density: 1.61 g/cm³ (20 °C)
MDL Number: MFCD00013814
CAS Number: 653-30-5
EINECS: 211-498-6
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR