α,α,α-Trifluoro-4-tolualdehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002181-25G
002181-100G
002181-250G
FLUO002181-25GEA
0
GBP
FLUO002181-25G
FLUO002181-100G
FLUO002181-250G
α,α,α-Trifluoro-4-tolualdehyde
α,α,α-Trifluoro-4-tolualdehyde
Formula:
C₈H₅F₃O |
MDL Number:
MFCD00006952 CAS Number: 455-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...