α,α,α-Trifluoro-3-tolualdehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001471-5G 001471-25G 001471-100G
FLUO001471-5GEA 0 GBP
FLUO001471-5G FLUO001471-25G FLUO001471-100G
α,α,α-Trifluoro-3-tolualdehyde
α,α,α-Trifluoro-3-tolualdehyde
Formula: C₈H₅F₃O
Boiling Pt: 83 °C (1013 hPa)
Density: 1,3024 g/cm³ (20 °C)
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR