α,α,α-Trifluoro-2-tolualdehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

007580-25G 007580-100G
FLUO007580-25GEA 0 GBP
FLUO007580-25G FLUO007580-100G
α,α,α-Trifluoro-2-tolualdehyde
α,α,α-Trifluoro-2-tolualdehyde
Formula: C₈H₅F₃O
Boiling Pt: 28…38 °C (0.4 mmHg)
Melting Pt: –40 °C
Density: 1,322 g/cm³ (20 °C)
MDL Number: MFCD00003337
CAS Number: 447-61-0
EINECS: 207-185-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR