4-(Trifluoromethyl)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005207-5G 005207-25G
FLUO005207-5GEA 0 GBP
FLUO005207-5G FLUO005207-25G
4-(Trifluoromethyl)phenylboronic acid
4-(Trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₂
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR