2-(Trifluoromethyl)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011012-1G 011012-5G
FLUO011012-1GEA 0 GBP
FLUO011012-1G FLUO011012-5G
2-(Trifluoromethyl)phenylboronic acid
2-(Trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₂
MDL Number: MFCD00236059
CAS Number: 1423-27-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR