2,3-Difluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005238-5G 005238.10G 005238-1G
FLUO005238-5GEA 0 GBP
FLUO005238-5G FLUO005238.10G FLUO005238-1G
2,3-Difluorotoluene
2,3-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 117…119 °C (1013 hPa)
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01631506
CAS Number: 3828-49-7
EINECS: 442-630-2
UN: 2388
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR