α,α,α,2,3-Pentafluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

005739-5G 005739-1G
FLUO005739-5GEA 0 GBP
FLUO005739-5G FLUO005739-1G
α,α,α,2,3-Pentafluorotoluene
α,α,α,2,3-Pentafluorotoluene
Formula: C₇H₃F₅
MDL Number: MFCD01631429
CAS Number: 64248-59-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR