m-Toluic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Methylbenzoic acid
M-Clarity™ quality level = MQ100
T36609-100G
T36609-500G
T36609-5G
SIALT36609-100GEA
64.9
GBP
SIALT36609-100G
SIALT36609-500G
SIALT36609-5G
m-Toluic acid, Sigma-Aldrich®
m-Toluic acid
Formula:
C₈H₈O₂ MW: 136.15 g/mol Boiling Pt: 263 °C (1013 hPa) Melting Pt: 108…111 °C Density: 1.054 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002523 CAS Number: 99-04-7 EINECS: 202-723-9 Merck Index: 12,09673 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...