4-tert-Butylbenzhydrazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-tert-Butylbenzoic hydrazide, 4-(tert-Butyl)benzohydrazide, 4-tert-Butylbenzene-1-carbohydrazide, 4-tert-Butylbenzoic acid hydrazide, 4-tert-Butylbenzohydrazide

CFF582.2.5 CFF582.5 018335-10G 018335-25G
CHMPCFF582.2.5EA 0 GBP
CHMPCFF582.2.5 CHMPCFF582.5 FLUO018335-10G FLUO018335-25G
4-tert-Butylbenzhydrazide
4-tert-Butylbenzhydrazide
Formula: C₁₁H₁₆N₂O
MDL Number: MFCD00014763
CAS Number: 43100-38-5
EINECS: 256-090-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR