3'-Hydroxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
328103-25G
328103-5G
SIAL328103-25GEA
46.2
GBP
SIAL328103-25G
SIAL328103-5G
3'-Hydroxyacetophenone, Sigma-Aldrich®
3'-Hydroxyacetophenone
Formula:
C₈H₈O₂ MW: 136.15 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 86…88 °C Density: 1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002298 CAS Number: 121-71-1 EINECS: 204-494-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...