2'-Hydroxyacetophenone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: o-Hydroxyacetophenone, 2-Acetophenol, 2-Hydroxyphenyl Methyl Ketone, 1-(2-Hydroxyphenyl)ethan-1-one, o-Acetylphenol, 1-(2-Hydroxyphenyl)-1-oxoethane, 1-Ethanoyl-2-hydroxybenzol, 2-Acetylphenol

M-Clarity™ quality level = MQ300

W354805-1KG W354805-SAMPLE
SIALW354805-1KGEA 127 GBP
SIALW354805-1KG SIALW354805-SAMPLE
2'-Hydroxyacetophenone, Sigma-Aldrich®
2'-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 213 °C (1013 hPa)
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002219
CAS Number: 118-93-4
EINECS: 204-288-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR