2'-Hydroxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
o-Hydroxyacetophenone, 2-Acetophenol, 2-Hydroxyphenyl Methyl Ketone, 1-(2-Hydroxyphenyl)ethan-1-one, o-Acetylphenol, 1-(2-Hydroxyphenyl)-1-oxoethane, 1-Ethanoyl-2-hydroxybenzol, 2-Acetylphenol
M-Clarity™ quality level = MQ100
H18607-100G
H18607-500G
H18607-5G
SIALH18607-100GEA
60.8
GBP
SIALH18607-100G
SIALH18607-500G
SIALH18607-5G
2'-Hydroxyacetophenone, Sigma-Aldrich®
2'-Hydroxyacetophenone
Formula:
C₈H₈O₂ MW: 136.15 g/mol Boiling Pt: 213 °C (1013 hPa) Density: 1.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002219 CAS Number: 118-93-4 EINECS: 204-288-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...