2',4'-Dimethoxyacetophenone

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2,4-Dimethoxyphenyl)ethan-1-one

D4099.25 D4099.100
XXXXD4099.25EA 0 GBP
XXXXD4099.25 XXXXD4099.100
2',4'-Dimethoxyacetophenone
2',4'-Dimethoxyacetophenone
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00008727
CAS Number: 829-20-9
EINECS: 212-587-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR