4-Methoxyphenylacetone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(p-Methoxyphenyl)-2-propanone, 1-(4-Methoxyphenyl)-2-propanone

M-Clarity™ quality level = MQ200

199176-10G 199176-50G
SIAL199176-10GEA 35.6 GBP
SIAL199176-10G SIAL199176-50G
4-Methoxyphenylacetone, Sigma-Aldrich®
4-Methoxyphenylacetone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 268
Melting Pt: –15 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008773
CAS Number: 122-84-9
EINECS: 204-578-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR