3'-Methoxyacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021712-50G 021712-250G
FLUO021712-50GEA 0 GBP
FLUO021712-50G FLUO021712-250G
3'-Methoxyacetophenone
3'-Methoxyacetophenone
Formula: C₉H₁₀O₂
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1,09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008736
CAS Number: 586-37-8
EINECS: 209-573-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR