ω-Methoxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
M9602-25G
M9602-5G
SIALM9602-25GEA
818
GBP
SIALM9602-25G
SIALM9602-5G
ω-Methoxyacetophenone, Sigma-Aldrich®
ω-Methoxyacetophenone
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 65 °C (0.5 mmHg) Density: 1.096 g/cm³ (18 °C) |
CAS Number:
4079-52-1 EINECS: 223-802-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...