3'-Aminoacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: m-Aminoacetophenone, 1-(3-Aminophenyl)ethanone

001722-100G 001722-500G
FLUO001722-100GEA 0 GBP
FLUO001722-100G FLUO001722-500G
3'-Aminoacetophenone
3'-Aminoacetophenone
Formula: C₈H₉NO
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00007796
CAS Number: 99-03-6
EINECS: 202-722-3
Merck Index: 13,00412

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR