Benzoyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018199-10G 018199-100G 018199-25G
FLUO018199-10GEA 0 GBP
FLUO018199-10G FLUO018199-100G FLUO018199-25G
Benzoyl isothiocyanate
Benzoyl isothiocyanate
Formula: C₈H₅NOS
MW: 163.2 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.197 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004815
CAS Number: 532-55-8
EINECS: 208-540-0
UN: 2810
ADR: 6.1,III
Merck Index: 13,01115

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR