2,6-Dichlorobenzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008820-1G
008820-10G
008820-25G
FLUO008820-1GEA
0
GBP
FLUO008820-1G
FLUO008820-10G
FLUO008820-25G
2,6-Dichlorobenzyl alcohol
2,6-Dichlorobenzyl alcohol
Formula:
C₇H₆Cl₂O Boiling Pt: 268 °C (1013 hPa) Melting Pt: 96…98 °C |
MDL Number:
MFCD00004609 CAS Number: 15258-73-8 EINECS: 239-300-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...