3-Chloro-o-toluidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

021716-250G 021716-1KG
FLUO021716-250GEA 0 GBP
FLUO021716-250G FLUO021716-1KG
3-Chloro-o-toluidine
3-Chloro-o-toluidine
Formula: C₇H₈ClN
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.185 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007766
CAS Number: 87-60-5
EINECS: 201-756-6
UN: 3429
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR