3,4-Dichlorobenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

013145-5G 013145-25G
FLUO013145-5GEA 0 GBP
FLUO013145-5G FLUO013145-25G
3,4-Dichlorobenzylamine
3,4-Dichlorobenzylamine
Formula: C₇H₇Cl₂N
Boiling Pt: 89…92 °C (0.5 mmHg)
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008114
CAS Number: 102-49-8
EINECS: 203-035-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR