4-Chloro-2-iodotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018385-10G 018385-25G
FLUO018385-10GEA 0 GBP
FLUO018385-10G FLUO018385-25G
4-Chloro-2-iodotoluene
4-Chloro-2-iodotoluene
Formula: C₇H₆ClI
MDL Number: MFCD00060661
CAS Number: 33184-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR