2-Chloro-4-fluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005623-1G 005623-5G
FLUO005623-1GEA 0 GBP
FLUO005623-1G FLUO005623-5G
2-Chloro-4-fluorobenzyl alcohol
2-Chloro-4-fluorobenzyl alcohol
Formula: C₇H₆ClFO
MDL Number: MFCD00236024
CAS Number: 208186-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR