4-Chloro-2-fluorobenzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009419-5G
009419-25G
FLUO009419-5GEA
0
GBP
FLUO009419-5G
FLUO009419-25G
4-Chloro-2-fluorobenzyl alcohol
4-Chloro-2-fluorobenzyl alcohol
Formula:
C₇H₆ClFO |
MDL Number:
MFCD00143285 CAS Number: 56456-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...