2-Chloro-6-fluorobenzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Chloro-6-fluorophenyl)methanol
004440-25G
004440-100G
FLUO004440-25GEA
0
GBP
FLUO004440-25G
FLUO004440-100G
2-Chloro-6-fluorobenzyl alcohol
2-Chloro-6-fluorobenzyl alcohol
Formula:
C₇H₆ClFO MW: 160.58 g/mol Boiling Pt: 232 Melting Pt: 40…41 °C |
MDL Number:
MFCD00004608 CAS Number: 56456-50-9 EINECS: 260-192-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...