3-Chloro-4-fluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005626-5G 005626-25G
FLUO005626-5GEA 0 GBP
FLUO005626-5G FLUO005626-25G
3-Chloro-4-fluorobenzyl alcohol
3-Chloro-4-fluorobenzyl alcohol
Formula: C₇H₆ClFO
MDL Number: MFCD01631550
CAS Number: 161446-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR