3-Chloro-2-fluorobenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005625-1G 005625-5G
FLUO005625-1GEA 0 GBP
FLUO005625-1G FLUO005625-5G
3-Chloro-2-fluorobenzyl alcohol
3-Chloro-2-fluorobenzyl alcohol
Formula: C₇H₆ClFO
MDL Number: MFCD01631549
CAS Number: 261723-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR